3-Methoxy-2-(trifluormethyl)benzylalkohol 98%

Lieferant: Apollo Scientific
Achtung

PC302404-1G PC302404-5G PC302404-10G
APOSPC302404-1GEA 148 EUR
APOSPC302404-1G APOSPC302404-5G APOSPC302404-10G
3-Methoxy-2-(trifluormethyl)benzylalkohol 98%
3-Methoxy-2-(trifluormethyl)benzylalkohol
Formel: C₉H₉F₃O₂
Molecular Weight: 206.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18398589
CAS-Nummer: 1261750-58-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR