4-Fluor-3-methyl-5-(trifluormethyl)benzylamin 98%

Lieferant: Apollo Scientific

PC302413-5G PC302413-1G
APOSPC302413-5GEA 339 EUR
APOSPC302413-5G APOSPC302413-1G
4-Fluor-3-methyl-5-(trifluormethyl)benzylamin 98%
4-Fluor-3-methyl-5-(trifluormethyl)benzylamin
Formel: C₉H₉F₄N
Molecular Weight: 207.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22201070
CAS-Nummer: 1373921-00-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR