2-Chloro-5-fluorophenylacetonitrile 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC302443-10G
PC302443-5G
PC302443-1G
APOSPC302443-10GEA
74.1
EUR
APOSPC302443-10G
APOSPC302443-5G
APOSPC302443-1G
2-Chloro-5-fluorophenylacetonitrile 97%
2-Chloro-5-fluorophenylacetonitrile
Formel:
C₈H₅ClFN Lagertemperatur: Raumtemperatur |
MDL:
MFCD06246270 CAS-Nummer: 395675-23-7 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...