2-Methyl-5-(trifluormethoxy)anisol 97%

Lieferant: Apollo Scientific

PC302471-1G PC302471-5G
APOSPC302471-1GEA 159 EUR
APOSPC302471-1G APOSPC302471-5G
2-Methyl-5-(trifluormethoxy)anisol 97%
2-Methyl-5-(trifluormethoxy)anisol
Formel: C₉H₉F₃O₂
Molecular Weight: 206.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18399698
CAS-Nummer: 1261444-86-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR