3-(3,5-Difluoro-4-methoxyphenyl)propionic acid 98%

Lieferant: Apollo Scientific

PC302479-1G PC302479-5G
APOSPC302479-1GEA 207 EUR
APOSPC302479-1G APOSPC302479-5G
3-(3,5-Difluoro-4-methoxyphenyl)propionic acid 98%
3-(3,5-Difluoro-4-methoxyphenyl)propionic acid
Formel: C₁₀H₁₀F₂O₃
Molecular Weight: 216,18 g/mol
Schmelzpunkt: 83…86 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09832373
CAS-Nummer: 105219-44-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR