3-(5-Chlor-2-(trifluormethoxy)phenyl)propionsäure 97%

Lieferant: Apollo Scientific

PC302517-1G PC302517-5G
APOSPC302517-1GEA 171 EUR
APOSPC302517-1G APOSPC302517-5G
3-(5-Chlor-2-(trifluormethoxy)phenyl)propionsäure 97%
3-(5-Chlor-2-(trifluormethoxy)phenyl)propionsäure
Formel: C₁₀H₈ClF₃O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11519362
CAS-Nummer: 1092461-23-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR