3-Chloro-4-ethoxy-5-fluorophenylacetonitrile 97%
Lieferant: Apollo Scientific
PC302529-1G
PC302529-5G
APOSPC302529-1GEA
159
EUR
APOSPC302529-1G
APOSPC302529-5G
3-Chloro-4-ethoxy-5-fluorophenylacetonitrile 97%
3-Chloro-4-ethoxy-5-fluorophenylacetonitrile
Formel:
C₁₀H₉ClFNO Molecular Weight: 213,64 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09832343 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...