3-Chlor-5-fluor-4-methoxybenzaldehyd 97%
Lieferant: Apollo Scientific
|
Achtung
|
PC302536-5G
PC302536-1G
APOSPC302536-5GEA
154
EUR
APOSPC302536-5G
APOSPC302536-1G
3-Chlor-5-fluor-4-methoxybenzaldehyd 97%
3-Chloro-5-fluoro-4-methoxybenzaldehyde
Formel:
C₈H₆ClFO₂ Molecular Weight: 188,59 g/mol Schmelzpunkt: 43…45 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115863 CAS-Nummer: 177994-49-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...