3-Chlor-5-fluor-4-methoxybenzylamin 97%

Lieferant: Apollo Scientific

PC302538-1G PC302538-5G PC302538-250MG
APOSPC302538-1GEA 183 EUR
APOSPC302538-1G APOSPC302538-5G APOSPC302538-250MG
3-Chlor-5-fluor-4-methoxybenzylamin 97%
3-Chlor-5-fluor-4-methoxybenzylamin
Formel: C₈H₉ClFNO
Molecular Weight: 189,62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07786699
CAS-Nummer: 887581-86-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR