(3-Ethoxy-2,6-difluorphenyl)essigsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC302547-1G
PC302547-5G
APOSPC302547-1GEA
116
EUR
APOSPC302547-1G
APOSPC302547-5G
(3-Ethoxy-2,6-difluorphenyl)essigsäure 98%
(3-Ethoxy-2,6-difluorphenyl)essigsäure
Formel:
C₁₀H₁₀F₂O₃ Molecular Weight: 216,18 g/mol Schmelzpunkt: 101…103 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD11519374 CAS-Nummer: 1092461-32-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...