3-Fluor-o-anisaldehyd 98%

Lieferant: Apollo Scientific
Achtung

PC302548-5G PC302548-1G
APOSPC302548-5GEA 139 EUR
APOSPC302548-5G APOSPC302548-1G
3-Fluor-o-anisaldehyd 98%
3-Fluor-o-anisaldehyd
Formel: C₈H₇FO₂
Molecular Weight: 154,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04112486
CAS-Nummer: 74266-68-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR