4-(Difluormethoxy)-2-fluorbenzylbromid 97%

Lieferant: Apollo Scientific
Gefahr

PC302561-1G PC302561-5G PC302561-10G
APOSPC302561-1GEA 115 EUR
APOSPC302561-1G APOSPC302561-5G APOSPC302561-10G
4-(Difluormethoxy)-2-fluorbenzylbromid 97%
4-(Difluormethoxy)-2-fluorbenzylbromid
Formel: C₈H₆BrF₃O
Molecular Weight: 255,03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09832360
CAS-Nummer: 1017779-33-7
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR