4-Chloro-2-fluoro-3-methoxybenzyl bromide

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-Chloro-2-fluoro-3-methoxybenzyl bromide

PC302571-1G PC302571-250MG PC302571-5G
APOSPC302571-1GEA 190 EUR
APOSPC302571-1G APOSPC302571-250MG APOSPC302571-5G
4-Chloro-2-fluoro-3-methoxybenzyl bromide
4-Chloro-2-fluoro-3-methoxybenzyl bromide
Formel: C₈H₇BrClFO
Molecular Weight: 253,50 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19687182
CAS-Nummer: 1323966-35-3
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR