4-Ethoxy-3-(trifluoromethyl)benzylamine 98%

Lieferant: Apollo Scientific

Synonyme: 4-ETHOXY-3-(TRIFLUOROMETHYL)BENZYLAMINE

PC302587-5G PC302587-1G
APOSPC302587-5GEA 185 EUR
APOSPC302587-5G APOSPC302587-1G
4-Ethoxy-3-(trifluoromethyl)benzylamine 98%
4-Ethoxy-3-(trifluoromethyl)benzylamine
Formel: C₁₀H₁₂F₃NO
Molecular Weight: 219.21 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13196584
CAS-Nummer: 1206593-27-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR