4-Fluoro-2-(trifluoromethoxy)benzamide

Lieferant: Apollo Scientific

PC302590-1G PC302590-5G
APOSPC302590-1GEA 0 EUR
APOSPC302590-1G APOSPC302590-5G
4-Fluoro-2-(trifluoromethoxy)benzamide
4-Fluoro-2-(trifluoromethoxy)benzamide
Formel: C₈H₅F₄NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18907236
CAS-Nummer: 1323966-41-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR