5-Fluoro-2-methoxyphenylacetonitrile 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-(5-Fluoro-2-methoxyphenyl)acetonitrile
PC302623-1G
PC302623-5G
PC302623-10G
APOSPC302623-1GEA
49.3
EUR
APOSPC302623-1G
APOSPC302623-5G
APOSPC302623-10G
5-Fluoro-2-methoxyphenylacetonitrile 98%
5-Fluoro-2-methoxyphenylacetonitrile
Formel:
C₉H₈FNO Molecular Weight: 165,17 g/mol Schmelzpunkt: 53…56 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00671773 CAS-Nummer: 501008-41-9 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...