6-Chlor-2-fluor-3-methoxybenzamid 97%

Lieferant: Apollo Scientific

PC302632-10G PC302632-5G PC302632-1G PC302632-25G PC302632-100G
APOSPC302632-10GEA 246 EUR
APOSPC302632-10G APOSPC302632-5G APOSPC302632-1G APOSPC302632-25G APOSPC302632-100G
6-Chlor-2-fluor-3-methoxybenzamid 97%
6-Chlor-2-fluor-3-methoxybenzamid
Formel: C₈H₇ClFNO₂
Molecular Weight: 203.60 g/mol
Schmelzpunkt: 143…145 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06660354
CAS-Nummer: 886761-75-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR