2-Chlor-3-(trifluormethyl)anisol 98%

Lieferant: Apollo Scientific

PC302638-10G PC302638-5G PC302638-1G
APOSPC302638-10GEA 246 EUR
APOSPC302638-10G APOSPC302638-5G APOSPC302638-1G
2-Chlor-3-(trifluormethyl)anisol 98%
2-Chlor-3-(trifluormethyl)anisol
Formel: C₈H₆ClF₃O
Molecular Weight: 210,58 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06660322
CAS-Nummer: 886503-84-0
UN: 3272
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR