4-Chlor-2-(trifluormethyl)benzylbromid 98%

Lieferant: Apollo Scientific
Gefahr

PC302644-5G PC302644-1G PC302644-25G
APOSPC302644-5GEA 159 EUR
APOSPC302644-5G APOSPC302644-1G APOSPC302644-25G
4-Chlor-2-(trifluormethyl)benzylbromid 98%
4-Chlor-2-(trifluormethyl)benzylbromid
Formel: C₈H₅BrClF₃
Molecular Weight: 273,48 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115840
CAS-Nummer: 886496-75-9
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR