4-Chlor-3-(trifluormethoxy)phenylessigsäure 97%
Lieferant: Apollo Scientific
Synonyme:
2-(4-Chloro-3-(trifluoromethoxy)phenyl)acetic acid
PC302645-1G
PC302645-5G
PC302645-10G
APOSPC302645-1GEA
50.9
EUR
APOSPC302645-1G
APOSPC302645-5G
APOSPC302645-10G
4-Chlor-3-(trifluormethoxy)phenylessigsäure 97%
4-Chlor-3-(trifluormethoxy)phenylessigsäure
Formel:
C₉H₆ClF₃O₃ Molecular Weight: 254,59 g/mol Schmelzpunkt: 61…63 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06660251 CAS-Nummer: 886501-02-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...