4-Ethoxy-3-(trifluoromethyl)phenol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(trifluoromethyl)phenetyl alcohol

PC302647-1G PC302647-5G
APOSPC302647-1GEA 180 EUR
APOSPC302647-1G APOSPC302647-5G
4-Ethoxy-3-(trifluoromethyl)phenol 98%
4-Ethoxy-3-(trifluoromethyl)phenol
Formel: C₉H₉F₃O₂
Molecular Weight: 206,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13196587
CAS-Nummer: 1206593-24-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR