2,3-Dichlor-6-(trifluormethyl)benzoesäure 98%

Lieferant: Apollo Scientific

PC302655-5G PC302655-1G
APOSPC302655-5GEA 116 EUR
APOSPC302655-5G APOSPC302655-1G
2,3-Dichlor-6-(trifluormethyl)benzoesäure 98%
2,3-Dichloro-6-(trifluoromethyl)benzoic acid
Formel: C₈H₃Cl₂F₃O₂
Molecular Weight: 259.01 g/mol
Schmelzpunkt: 94…96 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06660270
CAS-Nummer: 25922-43-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR