2,3-Dichlor-6-fluoranisol 98%

Lieferant: Apollo Scientific

PC302657-5G PC302657-1G
APOSPC302657-5GEA 224 EUR
APOSPC302657-5G APOSPC302657-1G
2,3-Dichlor-6-fluoranisol 98%
2,3-Dichlor-6-fluoranisol
Formel: C₇H₅Cl₂FO
Molecular Weight: 195,02 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09832260
CAS-Nummer: 1017777-48-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR