(4-Ethoxy-2,6-difluorphenyl)essigsäure 97%

Lieferant: Apollo Scientific

Synonyme: 2,6-Difluoro-4-ethoxyphenylacetic acid

PC302670-1G PC302670-5G
APOSPC302670-1GEA 159 EUR
APOSPC302670-1G APOSPC302670-5G
(4-Ethoxy-2,6-difluorphenyl)essigsäure 97%
(4-Ethoxy-2,6-difluorphenyl)essigsäure
Formel: C₁₀H₁₀F₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09258682
CAS-Nummer: 1017779-16-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR