2-Chlor-3,6-difluoranilin 98%
Lieferant: Apollo Scientific
PC302672-10G
PC302672-5G
PC302672-1G
APOSPC302672-10GEA
328
EUR
APOSPC302672-10G
APOSPC302672-5G
APOSPC302672-1G
2-Chlor-3,6-difluoranilin 98%
2-Chlor-3,6-difluoranilin
Formel:
C₆H₄ClF₂N Molecular Weight: 163,55 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06660176 CAS-Nummer: 287172-80-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...