2-Chlor-4-(trifluormethyl)benzaldehyd 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-4-(trifluoromethyl)-benzaldehyde

PC302673-1G PC302673-5G
APOSPC302673-1GEA 40.6 EUR
APOSPC302673-1G APOSPC302673-5G
2-Chlor-4-(trifluormethyl)benzaldehyd 97%
2-Chloro-4-(trifluoromethyl)benzaldehyde
Formel: C₈H₄ClF₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD04112538
CAS-Nummer: 82096-91-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR