3,5-Difluor-2-ethoxybenzonitril 98%
Lieferant: Apollo Scientific
Synonyme:
2-Ethoxy-3,5-difluorobenzonitrile
PC302679-10G
PC302679-5G
PC302679-1G
APOSPC302679-10GEA
282
EUR
APOSPC302679-10G
APOSPC302679-5G
APOSPC302679-1G
3,5-Difluor-2-ethoxybenzonitril 98%
3,5-Difluor-2-ethoxybenzonitril
Formel:
C₉H₇F₂NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD09258716 CAS-Nummer: 1017779-80-4 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...