2-Fluoro-4-methylbenzoyl chloride 97%

Lieferant: Apollo Scientific
Gefahr

PC302683-5G PC302683-1G
APOSPC302683-5GEA 522 EUR
APOSPC302683-5G APOSPC302683-1G
2-Fluoro-4-methylbenzoyl chloride 97%
2-Fluoro-4-methylbenzoyl chloride
Formel: C₈H₆ClFO
Molecular Weight: 172,59 g/mol
Schmelzpunkt: 43…47 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115879
CAS-Nummer: 59189-98-9
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR