2-Methoxy-4-(trifluormethoxy)benzylbromid 97%

Lieferant: Apollo Scientific
Gefahr

PC302690-10G PC302690-5G PC302690-1G
APOSPC302690-10GEA 440 EUR
APOSPC302690-10G APOSPC302690-5G APOSPC302690-1G
2-Methoxy-4-(trifluormethoxy)benzylbromid 97%
2-Methoxy-4-(trifluormethoxy)benzylbromid
Formel: C₉H₈BrF₃O₂
Molecular Weight: 285.06 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04115970
CAS-Nummer: 886500-35-2
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR