2-Methoxy-5-(trifluormethyl)benzylbromid

Lieferant: Apollo Scientific
Gefahr

PC302694-5G PC302694-1G PC302694-25G
APOSPC302694-5GEA 122 EUR
APOSPC302694-5G APOSPC302694-1G APOSPC302694-25G
2-Methoxy-5-(trifluormethyl)benzylbromid
2-Methoxy-5-(trifluormethyl)benzylbromid
Formel: C₉H₈BrF₃O
Molecular Weight: 269.06 g/mol
Schmelzpunkt: 34…37 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06660318
CAS-Nummer: 685126-88-9
UN: 3261
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR