3,5-Difluor-2-methylbenzamid 98%

Lieferant: Apollo Scientific

PC302700-1G PC302700-5G PC302700-10G
APOSPC302700-1GEA 136 EUR
APOSPC302700-1G APOSPC302700-5G APOSPC302700-10G
3,5-Difluor-2-methylbenzamid 98%
3,5-Difluor-2-methylbenzamid
Formel: C₈H₇F₂NO
Molecular Weight: 171,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19687171
CAS-Nummer: 1323966-37-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR