(3,6-Dichlor-2-fluorphenyl)essigsäure 98%

Lieferant: Apollo Scientific

PC302704-1G PC302704-5G PC302704-10G
APOSPC302704-1GEA 151 EUR
APOSPC302704-1G APOSPC302704-5G APOSPC302704-10G
(3,6-Dichlor-2-fluorphenyl)essigsäure 98%
(3,6-Dichlor-2-fluorphenyl)essigsäure
Formel: C₈H₅Cl₂FO₂
Molecular Weight: 223,03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09025391
CAS-Nummer: 916420-71-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR