3-Bromo-4-methoxy-5-(trifluoromethyl)aniline 98%

Lieferant: Apollo Scientific

Synonyme: 3-Bromo-4-methoxy-5-(trifluoromethyl)aniline

PC302709-1G PC302709-5G PC302709-10G
APOSPC302709-1GEA 136 EUR
APOSPC302709-1G APOSPC302709-5G APOSPC302709-10G
3-Bromo-4-methoxy-5-(trifluoromethyl)aniline 98%
3-Bromo-4-methoxy-5-(trifluoromethyl)aniline
Formel: C₈H₇BrF₃NO
Molecular Weight: 270,05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22201052
CAS-Nummer: 1373920-70-7
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR