3-Chlor-4-(trifluormethoxy)phenylessigsäure 97%

Lieferant: Apollo Scientific
Achtung

PC302712-1G PC302712-5G
APOSPC302712-1GEA 140 EUR
APOSPC302712-1G APOSPC302712-5G
3-Chlor-4-(trifluormethoxy)phenylessigsäure 97%
3-Chlor-4-(trifluormethoxy)phenylessigsäure
Formel: C₉H₆ClF₃O₃
Molecular Weight: 254.59 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09832392
CAS-Nummer: 1017779-77-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR