3-Ethoxy-2,4-difluorbenzaldehyd 98%

Lieferant: Apollo Scientific

PC302721-25G PC302721-5G PC302721-1G
APOSPC302721-25GEA 1160 EUR
APOSPC302721-25G APOSPC302721-5G APOSPC302721-1G
3-Ethoxy-2,4-difluorbenzaldehyd 98%
3-Ethoxy-2,4-difluorbenzaldehyd
Formel: C₉H₈F₂O₂
Molecular Weight: 186,16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09258722
CAS-Nummer: 1017779-87-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR