4-Chlor-2-(trifluormethoxy)benzylamin 98%
Lieferant: Apollo Scientific
PC302738-1G
PC302738-5G
APOSPC302738-1GEA
136
EUR
APOSPC302738-1G
APOSPC302738-5G
4-Chlor-2-(trifluormethoxy)benzylamin 98%
4-Chlor-2-(trifluormethoxy)benzylamin
Formel:
C₈H₇ClF₃NO Molecular Weight: 225.60 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18393826 CAS-Nummer: 1261605-72-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...