4-Chlor-3-(trifluormethoxy)benzaldehyd 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC302740-10G
PC302740-5G
PC302740-1G
APOSPC302740-10GEA
315
EUR
APOSPC302740-10G
APOSPC302740-5G
APOSPC302740-1G
4-Chlor-3-(trifluormethoxy)benzaldehyd 98%
4-Chloro-3-(trifluoromethoxy)benzaldehyde
Formel:
C₈H₄ClF₃O₂ Molecular Weight: 224,57 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06660219 CAS-Nummer: 886499-59-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...