4'-Ethoxy-2',3'-difluoracetophenon 97%
Lieferant: Apollo Scientific
PC302744-10G
PC302744-5G
PC302744-1G
APOSPC302744-10GEA
282
EUR
APOSPC302744-10G
APOSPC302744-5G
APOSPC302744-1G
4'-Ethoxy-2',3'-difluoracetophenon 97%
4'-Ethoxy-2',3'-difluoracetophenon
Formel:
C₁₀H₁₀F₂O₂ Schmelzpunkt: 39…42 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09258710 CAS-Nummer: 1017779-68-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...