4-Ethoxy-2,3-difluorbenzylalkohol 98%
Lieferant: Apollo Scientific
|
Achtung
|
PC302745-10G
APOSPC302745-10GEA
0
EUR
APOSPC302745-10G
4-Ethoxy-2,3-difluorbenzylalkohol 98%
4-Ethoxy-2,3-difluorbenzylalkohol
Formel:
C₉H₁₀F₂O₂ Molecular Weight: 188,17 g/mol Schmelzpunkt: 52…56 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09258704 CAS-Nummer: 293299-94-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...