4-Ethoxy-2,6-difluorbenzaldehyd

Lieferant: Apollo Scientific
Achtung

PC302746-1G PC302746-5G PC302746-25G
APOSPC302746-1GEA 58 EUR
APOSPC302746-1G APOSPC302746-5G APOSPC302746-25G
4-Ethoxy-2,6-difluorbenzaldehyd
4-Ethoxy-2,6-difluorbenzaldehyd
Formel: C₉H₈F₂O₂
Molecular Weight: 186,16 g/mol
Schmelzpunkt: 70…72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09258695
CAS-Nummer: 1017779-48-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR