4-Methyl-3-(trifluormethyl)phenol 98%
Lieferant: Apollo Scientific
|
Gefahr
|
PC302756-1G
PC302756-5G
PC302756-10G
APOSPC302756-1GEA
203
EUR
APOSPC302756-1G
APOSPC302756-5G
APOSPC302756-10G
4-Methyl-3-(trifluormethyl)phenol 98%
4-Methyl-3-(trifluormethyl)phenol
Formel:
C₈H₇F₃O Molecular Weight: 176.14 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09832294 CAS-Nummer: 1017778-16-3 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...