5-Fluor-2-(trifluormethoxy)anilin 98%

Lieferant: Apollo Scientific

PC302761-1G PC302761-5G PC302761-10G
APOSPC302761-1GEA 0 EUR
APOSPC302761-1G APOSPC302761-5G APOSPC302761-10G
5-Fluor-2-(trifluormethoxy)anilin 98%
5-Fluor-2-(trifluormethoxy)anilin
Formel: C₇H₅F₄NO
Lagertemperatur: Raumtemperatur
MDL: MFCD16652514
CAS-Nummer: 123572-63-4
UN: 2810
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR