2-Chlor-6-fluor-3-methoxybenzonitril 98%
Lieferant: Apollo Scientific
|
Gefahr
|
PC302764-10G
PC302764-5G
PC302764-1G
APOSPC302764-10GEA
386
EUR
APOSPC302764-10G
APOSPC302764-5G
APOSPC302764-1G
2-Chlor-6-fluor-3-methoxybenzonitril 98%
2-Chlor-6-fluor-3-methoxybenzonitril
Formel:
C₈H₅ClFNO Schmelzpunkt: 100…104 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06660326 CAS-Nummer: 886761-59-7 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...