4-Chlor-2-fluorbenzylamin 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
(4-Chloro-2-fluorophenyl)methanamine
PC302779-5G
PC302779-1G
APOSPC302779-5GEA
68.3
EUR
APOSPC302779-5G
APOSPC302779-1G
4-Chlor-2-fluorbenzylamin 98%
4-Chlor-2-fluorbenzylamin
Formel:
C₇H₇ClFN Molecular Weight: 159.59 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06212852 CAS-Nummer: 72235-57-5 UN: 2735 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...