2,3-Difluor-6-methoxybenzylbromid 97%
Lieferant: Apollo Scientific
|
Gefahr
|
PC302788-10G
PC302788-5G
PC302788-1G
APOSPC302788-10GEA
348
EUR
APOSPC302788-10G
APOSPC302788-5G
APOSPC302788-1G
2,3-Difluor-6-methoxybenzylbromid 97%
2,3-Difluor-6-methoxybenzylbromid
Formel:
C₈H₇BrF₂O Schmelzpunkt: 46…48 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04116023 CAS-Nummer: 886501-83-3 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...