2,6-Difluor-3-methoxybenzylbromid 98%

Lieferant: Apollo Scientific
Gefahr

PC302791-10G PC302791-5G PC302791-1G
APOSPC302791-10GEA 328 EUR
APOSPC302791-10G APOSPC302791-5G APOSPC302791-1G
2,6-Difluor-3-methoxybenzylbromid 98%
2,6-Difluor-3-methoxybenzylbromid
Formel: C₈H₇BrF₂O
Molecular Weight: 237,04 g/mol
Schmelzpunkt: 53…56 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04115906
CAS-Nummer: 886498-55-1
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR