2-Chloro-5-(pentafluorothio)benzamide 98%

Lieferant: Apollo Scientific

Synonyme: 2-Chloro-5-(pentafluorosulphur)benzamide

PC302794-1G PC302794-5G PC302794-250MG
APOSPC302794-1GEA 342 EUR
APOSPC302794-1G APOSPC302794-5G APOSPC302794-250MG
2-Chloro-5-(pentafluorothio)benzamide 98%
2-Chloro-5-(pentafluorothio)benzamide
Formel: C₇H₅ClF₅NOS
Molecular Weight: 281,63 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22988934
CAS-Nummer: 1431329-71-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR