2-Fluor-4-methoxybenzamid 97%

Lieferant: Apollo Scientific

PC302803-10G PC302803-5G PC302803-1G
APOSPC302803-10GEA 325 EUR
APOSPC302803-10G APOSPC302803-5G APOSPC302803-1G
2-Fluor-4-methoxybenzamid 97%
2-Fluor-4-methoxybenzamid
Formel: C₈H₈FNO₂
Molecular Weight: 169,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01117440
CAS-Nummer: 874804-31-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR