2-Fluoro-5-methoxy-3-(trifluoromethyl)benzamide 97%

Lieferant: Apollo Scientific

Synonyme: 2-Fluoro-5-methoxy-3-(trifluoromethyl)benzamide

PC302804-5G PC302804-1G
APOSPC302804-5GEA 848 EUR
APOSPC302804-5G APOSPC302804-1G
2-Fluoro-5-methoxy-3-(trifluoromethyl)benzamide 97%
2-Fluoro-5-methoxy-3-(trifluoromethyl)benzamide
Formel: C₉H₇F₄NO₂
Molecular Weight: 237.15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22201042
CAS-Nummer: 1373920-76-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR