α-Brom-3-chlor-2,4-difluortoluol 97%

Lieferant: Apollo Scientific
Gefahr

PC302827-10G PC302827-5G PC302827-1G PC302827-25G PC302827-100G
APOSPC302827-10GEA 513 EUR
APOSPC302827-10G APOSPC302827-5G APOSPC302827-1G APOSPC302827-25G APOSPC302827-100G
α-Brom-3-chlor-2,4-difluortoluol 97%
α-Brom-3-chlor-2,4-difluortoluol
Formel: C₇H₄BrClF₂
Molecular Weight: 241.46 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04116003
CAS-Nummer: 886501-15-1
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR